Anti-PRKAR1B Rabbit Polyclonal Antibody

Lieferant: Abgent
AP7051A
ABGEAP7051AEA 354 EUR
ABGEAP7051A
Anti-PRKAR1B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRKAR1B (protein kinase, cAMP-dependent, regulatory, type I, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR