Anti-CDKL2 Rabbit Polyclonal Antibody
Lieferant: Abgent
AP7122A
ABGEAP7122AEA 354 EUR
ABGEAP7122A
Anti-CDKL2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDKL2 (cyclin-dependent kinase-like 2 (CDC2-related kinase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human