Anti-EIF2AK4 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP7130A
ABGEAP7130AEA 354 EUR
ABGEAP7130A
Anti-EIF2AK4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EIF2AK4 (eukaryotic translation initiation factor 2 alpha kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR