Anti-PFKM Rabbit Polyclonal Antibody

Lieferant: Abgent
AP8137A
ABGEAP8137AEA 354 EUR
ABGEAP8137A
Anti-PFKM Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PFKM (phosphofructokinase, muscle)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR