Anti-ADRBK2 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP9092A
ABGEAP9092AEA 354 EUR
ABGEAP9092A
Anti-ADRBK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ADRBK2 (adrenergic, beta, receptor kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR