Anti-EIF4A2 Rabbit Polyclonal Antibody

Lieferant: Abgent
AP9190B
ABGEAP9190BEA 354 EUR
ABGEAP9190B
Anti-EIF4A2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EIF4A2 (eukaryotic translation initiation factor 4A2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR