LOX assay kit (fluorometric)
Lieferant: Abnova
KA4191
ABNOKA4191EA
503
EUR
ABNOKA4191
LOX assay kit (fluorometric)
Tests
Enzymtests
LOX assay kit (fluorometric) is designed for the measurement of LOX.
- Suitable sample: biological samples
- Sample volume: 50 µl
- Calibration range: 0,04 to 4 µg/ml
Lieferumfang: The kit includes component A: amplite HRP substrate (light sensitive) (1 vial), component B: assay buffer (50 ml), component C: horseradish peroxidase (50 units) (1 vial), component D: DMSO (200 μl).
Bitte beachten For research use only (RUO). Store the kit at –20 °C and avoid from light.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...