TBARS ASSAY KIT TBARS ASSAY KIT IS USED FOR THE QUANTITATIVE DETERMINATION OF THIOBARBITURIC ACID REACTIVE SUBSTANCES THROUGH THE D ETECTION OF AN ADDUCT OF MALONDIALDEHYDE (MDA) AND THIOBARBITURIC ACID (TBA) 1 Kit 1 * 1 KIT
Lieferant: Abnova
TBARS ASSAY KIT TBARS ASSAY KIT IS USED FOR THE QUANTITATIVE DETERMINATION OF THIOBARBITURIC ACID REACTIVE SUBSTANCES THROUGH THE D ETECTION OF AN ADDUCT OF MALONDIALDEHYDE (MDA) AND THIOBARBITURIC ACID (TBA) 1 Kit 1 * 1 KIT