BSA I Restriction Endonuclease
Lieferant: Abnova
P9388.250UL
P9387.50UL
ABNOP9388.250ULEA
711
EUR
ABNOP9388.250UL
ABNOP9387.50UL
BSA I Restriction Endonuclease
Nukleinsäure-Reagenzien
Restriktionsenzym
The BSA I restriction endonuclease is a restriction endonuclease BsaI that can recognize specific sites expressed by Escherichia coli recombinantly. Restriction endonucleases are widely used in various fields such as gene positioning and cloning. This enzyme cleaves DNA rapidly for efficient gene linearization.