Wasser RNase_free

Lieferant: Abnova

Synonyme: Wasserstoffoxid, Hydrogenoxid

R0013-3 R0013-2
ABNOR0013-3EA 171 EUR
ABNOR0013-3 ABNOR0013-2
Wasser RNase_free
Wasser

RNase-free water are prepared from highly purified water by ultrafiltration without DEPC treatment.


  • Free from RNase and DNase
  • Ideal in RNA research

Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100
Schmelzpunkt: 0
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
Merck Index: 13,10098

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR