4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI) 1500 ng/ml Gegenfärbung

Lieferant: Abnova

Synonyme: 2-(4-Amidinophenyl)indole-6-carboxamidine Dihydrochloride, 2-(4-Carbamimidoylphenyl)-1H-indole-6-carboximidamide dihydrochloride, DAPI.2HCl, 2-(4-Carbamimidoylphenyl)indole-6-carboximidamide Dihydrochloride, DAPI 2HCl

U0031
ABNOU0031EA 196 EUR
ABNOU0031
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI) 1500 ng/ml Gegenfärbung
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI)

DAPI counterstain provides more distinct banding patterns in applications such as staining of paraffin sections nuclear DNA.


  • for Fluorescent In Situ Hybridization (Cell)


Recommended counterstain when a more intense DAPI stain is desired.


DAPI is the recommended counterstain for FISH probes. This concentration of DAPI provides a weaker DAPI stain usually preferred with smaller probe signals such as FISH probes.


Complementary reagent: Antifade solution (U0032) is comprised of antifade and glycerol in an aqueous buffer solution.

Formel: C₁₆H₁₅N₅·2HCl
Lagertemperatur: Kühlschrank
MDL: MFCD00012681
CAS-Nummer: 28718-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR