FLUORESCENT DYE 350-I PHALLOIDIN FLUORESCENCE DYE 350-I WITH PHALLOIDIN CONJUGATE. PROTEIN CONJUGATES PREPARED WITH FLUORESCENCE DY E 350-I ARE BRIGHT WITH FLUORESCENCE UNAFFECTED BY PH IN THE PHYSIOLOGICAL RANGE (PH 4-10) 300 Reactions 1 * 300 Assays
Lieferant: Abnova
FLUORESCENT DYE 350-I PHALLOIDIN FLUORESCENCE DYE 350-I WITH PHALLOIDIN CONJUGATE. PROTEIN CONJUGATES PREPARED WITH FLUORESCENCE DY E 350-I ARE BRIGHT WITH FLUORESCENCE UNAFFECTED BY PH IN THE PHYSIOLOGICAL RANGE (PH 4-10) 300 Reactions 1 * 300 Assays