Midget bubbler,Complete with 0-25mL (5mL divisions) graduated bottle and #7 Ace-Safe stopper with porosity B (70-100 micron) filter tip 1 * 1 ST
Lieferant: Ace Glass
Midget bubbler,Complete with 0-25mL (5mL divisions) graduated bottle and #7 Ace-Safe stopper with porosity B (70-100 micron) filter tip 1 * 1 ST