Cynomolgus Rhesus macaque CD47, Fc Tag (CD7-C5252) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 141 (Acc ession F7A802-1). In the region Gln 19 - Glu 141, the AA sequence of Cynomolgus and Rhesus macaque CD47 are homologus 1 * 100 µG
Lieferant: ACROBIOSYSTEMS
Cynomolgus Rhesus macaque CD47, Fc Tag (CD7-C5252) is expressed from human 293 cells (HEK293). It contains AA Gln 19 - Glu 141 (Acc ession F7A802-1). In the region Gln 19 - Glu 141, the AA sequence of Cynomolgus and Rhesus macaque CD47 are homologus 1 * 100 µG