Human CD27 Ligand, His,Flag Tag (active trimer) (MALS verified) (CDL-H52Da) is expressed from human 293 cells (HEK293). It contains AA Ser 52 - Pro 193 (Accession P32970-1) trimer Design. 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human CD27 Ligand, His,Flag Tag (active trimer) (MALS verified) (CDL-H52Da) is expressed from human 293 cells (HEK293). It contains AA Ser 52 - Pro 193 (Accession P32970-1) trimer Design. 1 * 1 mg