Human recombinant CD40 ligand (from HEK293 cells)
Lieferant: ACRO Biosystems
CDL-H52DB-1MG
ACRBCDL-H52DB-1MGEA 2645 EUR
ACRBCDL-H52DB-1MG
Human recombinant CD40 ligand (from HEK293 cells)
Proteine und Peptide
Human CD40 Ligand / TNFSF5 protein, His,Flag Tag (active trimer) (MALS verified).