Canine M-CSF R, His Tag (CSR-C82H3) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 510 (Accession E2RJV2-1 ). 1 * 100 µG
Lieferant: ACROBIOSYSTEMS
Canine M-CSF R, His Tag (CSR-C82H3) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 510 (Accession E2RJV2-1 ). 1 * 100 µG