Cynomolgus Rhesus macaque FGL1, Fc Tag (FG1-C5269) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Acc ession G7N0K6-1). 1 * 100 µG
Lieferant: ACROBIOSYSTEMS
Cynomolgus Rhesus macaque FGL1, Fc Tag (FG1-C5269) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Ile 312 (Acc ession G7N0K6-1). 1 * 100 µG