Human Her2, His Tag (HE2-H5225) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession P04626-1). 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Human Her2, His Tag (HE2-H5225) is expressed from human 293 cells (HEK293). It contains AA Thr 23 - Thr 652 (Accession P04626-1). 1 * 1 mg