Human PD-L2, Mouse IgG1 Fc Tag (PD2-H52A5) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accession A AI13679). 1 * 100 µG
Lieferant: ACROBIOSYSTEMS
Human PD-L2, Mouse IgG1 Fc Tag (PD2-H52A5) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accession A AI13679). 1 * 100 µG