Human PLGF, His Tag (PGF-H5229) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 170 (Accession NP002623.2). 1 * 20 µG
Lieferant: ACROBIOSYSTEMS
Human PLGF, His Tag (PGF-H5229) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Arg 170 (Accession NP002623.2). 1 * 20 µG