Human recombinant PLAU (from HEK293 cells)
Lieferant: ACRO Biosystems
PLU-H5228-1MG
PLU-H5228-50UG
ACRBPLU-H5228-1MGEA
2750
EUR
ACRBPLU-H5228-1MG
ACRBPLU-H5228-50UG
Human recombinant PLAU (from HEK293 cells)
Proteine und Peptide
Human PLAU / uPA Protein, His Tag (activated by trypsin) (active enzyme).