Recombinat Protein,Rhesus macaque Siglec-2 CD22 Protein, His Tag,Rhesus macaque Siglec-2, His Tag (SI2-R52Ha) is expressed from hum an 293 cells (HEK293). It contains AA Ser 20 - Arg 686 (Accession EHH29920.1).This protein carries a polyhistidine ta 1 * 1 mg
Lieferant: ACROBIOSYSTEMS
Recombinat Protein,Rhesus macaque Siglec-2 CD22 Protein, His Tag,Rhesus macaque Siglec-2, His Tag (SI2-R52Ha) is expressed from hum an 293 cells (HEK293). It contains AA Ser 20 - Arg 686 (Accession EHH29920.1).This protein carries a polyhistidine ta 1 * 1 mg