Paracetamol 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4'-Hydroxyacetanilid, 4-Acetamidophenol, Acetaminophen, N-(4-Hydroxyphenyl)acetamid, Paracetamolum, APAP, Ethansäure(4-hydroxyphenyl)amid, N-Acetyl-4-aminophenol
102330010
102330050
102332500
102330051
ACRO102330010EA
114
EUR
ACRO102330010
ACRO102330050
ACRO102332500
ACRO102330051
Paracetamol 98%
Paracetamol
Formel:
C₈H₉NO₂ Siedepunkt: 388 °C (1013 hPa) Schmelzpunkt: 168…172 °C Dichte: 1,293 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002328 CAS-Nummer: 103-90-2 EINECS: 203-157-5 Merck Index: 13,00048 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 168 to 172 °C |
HPLC | ≥97.5 % |
Gel strength | 500 to 1000 g/cm² (1.5 % at 20 °C) |
Ash | ≤6 % |
Microbial contamination | Absent (Salmonella) |
pH | 5.7 to 7.5 (1.5 % in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...