Acetophenon 98%, rein

Lieferant: Thermo Fisher Scientific
Achtung

102410010 102410025 102410050 102412500 102410100
ACRO102410010EA 45.3 EUR
ACRO102410010 ACRO102410025 ACRO102410050 ACRO102412500 ACRO102410100
Acetophenon 98%, rein
Formel: C₈H₈O
Siedepunkt: 202 °C (760 mmHg)
Dichte: 1,028 g/cm³
Flammpunkt: 77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008724
CAS-Nummer: 98-86-2
EINECS: 202-708-7
ADR: 9,
Merck Index: 13,00074

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5320 to 1.5350 (20°C, 589 nm)
GC ≥97.5 %
Visual transition interval from pH 9.4 (orange-brown) to pH 12.0 (violet)
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR