Aluminon

Lieferant: Thermo Fisher Scientific
Achtung

103060250 103061000
ACRO103060250EA 56.5 EUR
ACRO103060250 ACRO103061000
Aluminon
Aluminon
Formel: C₂₂H₂₃N₃O₉
Schmelzpunkt: 220…225 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00040925
CAS-Nummer: 569-58-4
EINECS: 209-319-1

Bestellen

Spezifikation Testergebnisse

Appearance Red to yellow-brown to red-brown Crystalline powder
Infrared spectrum Conforms
Sulfated ash ≤0.2 %
Solubility (1.0 % in water) Clear
Suitability (for Al determination) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR