2-Aminoacetophenon Hydrochlorid 96%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Phenacylamine hydrochloride
103100010
ACRO103100010EA
32.1
EUR
ACRO103100010
2-Aminoacetophenon Hydrochlorid 96%
2-Aminoacetophenon Hydrochlorid
Formel:
C₈H₁₀ClNO Molecular Weight: 171,63 g/mol Schmelzpunkt: 194 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012873 CAS-Nummer: 5468-37-1 |
Spezifikation Testergebnisse
Appearance | White to yellow to beige to light brown Powder or crystals |
Infrared spectrum | Conforms |
Titration Argentometric | 95.0 to 105.0 % |
HPLC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...