Ethyl-3-amino-1H-pyrazol-4-carboxylat 99%

Lieferant: Thermo Fisher Scientific

Synonyme: 3-Amino-4-carbethoxypyrazole, Ethyl 3-amino-1H-pyrazole-4-carboxylate

103310050
ACRO103310050EA 43.3 EUR
ACRO103310050
Ethyl-3-amino-1H-pyrazol-4-carboxylat 99%
Ethyl-3-amino-1H-pyrazol-4-carboxylat
Formel: C₆H₉N₃O₂
Molecular Weight: 155.16 g/mol
Schmelzpunkt: 103…107 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005238
CAS-Nummer: 6994-25-8
EINECS: 230-262-3

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 103 to 107 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR