2-Amino-5-chlorpyridin 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Chloropyridin-2-amine, 5-Chloro-2-pyridylamine

103410250 103411000 103415000
ACRO103410250EA 41.1 EUR
ACRO103410250 ACRO103411000 ACRO103415000
2-Amino-5-chlorpyridin 98%
2-Amino-5-chlorpyridin
Formel: C₅H₅ClN₂
Siedepunkt: 127…128 °C (11 mmHg)
Schmelzpunkt: 134…138 °C
Dichte: 1,514 g/cm³ (20 °C)
Flammpunkt: 160 °C
MDL: MFCD00006324
CAS-Nummer: 1072-98-6
EINECS: 214-020-4

Bestellen

Spezifikation Testergebnisse

Appearance Beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 134 to 138 °C
HPLC ≥97.5 %
Titration with HClO4 ≥97.5 %
Copper (Cu) ≤20 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤20 ppm
Silver (Ag) ≤10 ppm
Insoluble matter ≤0.005 % (in HNO3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR