2-Amino-4,6-dimethylpyridin 99%
Lieferant: Thermo Scientific
|
Achtung
|
103580050
ACRO103580050EA
41.8
EUR
ACRO103580050
2-Amino-4,6-dimethylpyridin 99%
2-Amino-4,6-dimethylpyridine
Formel:
C₇H₁₀N₂ Molecular Weight: 122,1700 g/mol Siedepunkt: 235 °C (760 mmHg) Schmelzpunkt: 65…69,5 °C Flammpunkt: 121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006322 CAS-Nummer: 5407-87-4 |
Spezifikation Testergebnisse
Appearance | White to brown Crystalline powder or solid |
Infrared spectrum | Conforms |
Melting point | 64 to 70 °C |
GC | ≥98.5 % |
Heavy metals (as Pb) | ≤2 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...