2-Amino-4,6-dimethylpyrimidin 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4,6-Dimethylpyrimidin-2-ylamin, 4,6-Dimethylpyrimidin-2-amin

103590250
ACRO103590250EA 65.4 EUR
ACRO103590250
2-Amino-4,6-dimethylpyrimidin 98%
2-Amino-4,6-dimethylpyrimidine
Formel: C₆H₉N₃
Molecular Weight: 123,16 g/mol
Schmelzpunkt: 150…154 °C
MDL: MFCD00006102
CAS-Nummer: 767-15-7
EINECS: 212-181-5

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 150 to 154 °C
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR