2-Amino-4-methylpyrimidin 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Methylpyrimidin-2-ylamin

104090100 104091000
ACRO104090100EA 24 EUR
ACRO104090100 ACRO104091000
2-Amino-4-methylpyrimidin 97%
2-Amino-4-methylpyrimidine
Formel: C₅H₇N₃
Molecular Weight: 109.13 g/mol
Siedepunkt: 283
Schmelzpunkt: 155…160 °C
MDL: MFCD00006101
CAS-Nummer: 108-52-1
EINECS: 203-591-5

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 155 to 160 °C
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR