2-Amino-1-phenylethanol 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: alpha-(Aminomethyl)benzyl Alcohol, DL-β-Hydroxyphenethylamine, Phenylethanolamine

104320100
ACRO104320100EA 94.1 EUR
ACRO104320100
2-Amino-1-phenylethanol 98%
2-Amino-1-phenylethanol

Appearance: Clear yellow Viscous liquid

Formel: C₈H₁₁NO
Molecular Weight: 137,18 g/mol
Siedepunkt: 160 °C (17 mmHg)
Schmelzpunkt: 56…60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008137
CAS-Nummer: 7568-93-6
UN: 3259
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 52 to 60 °C
HPLC ≥97.5 %
Specific optical rotation +200° to +220° (20 °C, 589 nm) (c=3, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR