2-Amino-3-methylpyridin 96%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Methyl-2-pyridylamin, 3-Methylpyridin-2-amin, 2-Amino-3-picolin
104380050
104381000
104385000
ACRO104380050EA
24
EUR
ACRO104380050
ACRO104381000
ACRO104385000
2-Amino-3-methylpyridin 96%
2-Amino-3-methylpyridin
Formel:
C₆H₈N₂ Molecular Weight: 108.14 g/mol Siedepunkt: 221,1 Schmelzpunkt: 32…34 °C Flammpunkt: 111 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006320 CAS-Nummer: 1603-40-3 EINECS: 216-501-4 UN: 2811 ADR: 6.1 III |
Spezifikation Testergebnisse
Appearance | Clear yellow Liquid after melting |
Infrared spectrum | Conforms |
Refractive index | 1.5813 to 1.5833 (20°C, 589 nm) |
Water | ≤0.8 % |
GC | ≥95 % |
Fluoride (F) | ≤10 ppm |
Arsenic (As) | ≤3 ppm |
Iron (Fe) | ≤50 ppm |
Lead (Pb) | ≤5 ppm |
Heavy metals (as Pb) | ≤10 ppm |
Insoluble matter | ≤0.3 % (in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...