2-Amino-3-methylpyridin 96%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3-Methyl-2-pyridylamin, 3-Methylpyridin-2-amin, 2-Amino-3-picolin

104380050 104381000 104385000
ACRO104380050EA 24 EUR
ACRO104380050 ACRO104381000 ACRO104385000
2-Amino-3-methylpyridin 96%
2-Amino-3-methylpyridin
Formel: C₆H₈N₂
Molecular Weight: 108.14 g/mol
Siedepunkt: 221,1
Schmelzpunkt: 32…34 °C
Flammpunkt: 111 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006320
CAS-Nummer: 1603-40-3
EINECS: 216-501-4
UN: 2811
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance Clear yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5813 to 1.5833 (20°C, 589 nm)
Water ≤0.8 %
GC ≥95 %
Fluoride (F) ≤10 ppm
Arsenic (As) ≤3 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤5 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.3 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR