2-Amino-6-methylpyridin 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
6-Methylpyridin-2-amin, 2-Amino-6-picolin, 6-Methyl-2-pyridylamin
104400050
104401000
104405000
ACRO104400050EA
23.7
EUR
ACRO104400050
ACRO104401000
ACRO104405000
2-Amino-6-methylpyridin 98%
2-Amino-6-methylpyridin
Formel:
C₆H₈N₂ Molecular Weight: 108.14 g/mol Siedepunkt: 208…209 °C (760 mmHg) Schmelzpunkt: 40…45 °C Flammpunkt: 103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006331 CAS-Nummer: 1824-81-3 EINECS: 217-360-1 UN: 2811 ADR: 6.1 II |
Spezifikation Testergebnisse
Appearance | Colorless to white to yellow to light orange Low melting crystalline solid |
Infrared spectrum | Conforms |
Melting point | 40 to 45 °C |
GC | ≥97.5 % |
Particle size | ≤0.4 % (dust) |
pH | 3.5 to 6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...