2-Aminopyrimidin 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Pyrimidin-2-ylamin
104580050
104581000
104585000
ACRO104580050EA
22.4
EUR
ACRO104580050
ACRO104581000
ACRO104585000
2-Aminopyrimidin 98%
2-Aminopyrimidin
Formel:
C₄H₅N₃ Molecular Weight: 95.1 g/mol Siedepunkt: 158 °C (186mbar) Schmelzpunkt: 124…127 °C Flammpunkt: 103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006089 CAS-Nummer: 109-12-6 EINECS: 203-648-4 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Melting point | 123 to 127 °C |
Titration with HClO4 | ≥97.5 % |
BSC Certification | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...