11-Aminoundecansäure 97%
Lieferant: Thermo Fisher Scientific
104710050
104711000
104712500
ACRO104710050EA
30.4
EUR
ACRO104710050
ACRO104711000
ACRO104712500
11-Aminoundecansäure 97%
11-Aminoundecanoic acid
Formel:
C₁₁H₂₃NO₂ Siedepunkt: 331 °C (1013 hPa) Schmelzpunkt: 187…190 °C |
MDL:
MFCD00008150 CAS-Nummer: 2432-99-7 EINECS: 219-417-6 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 187 to 190 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...