D-Amygdalin hydrate 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: D-Mandelonitrile-beta-D-glucosido-6-beta-D-glucoside hydrate, Amygdalin hydrate

104730050
ACRO104730050EA 86.4 EUR
ACRO104730050
D-Amygdalin hydrate 95%
D-Amygdalin hydrate

CAS No.: 29883-15-6


Appearance: White Crystalline powder

Formel: C₂₀H₂₇NO₁₁·xH₂O
Molecular Weight: 457,44 g/mol (anhy)
Schmelzpunkt: 223…226 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006598
CAS-Nummer: 29883-15-6

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤5 % (K.F.)
Sulfated ash ≤0.1 % (on dry basis)
Solubility (4 % in water) Clear
HPLC ≥94 % (ELSD)
Specific optical rotation -36.5° to -40.5° (20°C, 589 nm) (c=4, H2O) (on dry basis)
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Stabilizer 0.5 to 1.0 % ethanol
Residue after evaporation ≤0.001 %
Acetone and aldehyde ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR