Benzethoniumchlorid 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Hyamine® 1622
105381000
105385000
105380025
ACRO105381000EA
67.7
EUR
ACRO105381000
ACRO105385000
ACRO105380025
Benzethoniumchlorid 97%
Benzethoniumchlorid
Formel:
C₂₇H₄₂ClNO₂ Schmelzpunkt: 158…163 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011742 CAS-Nummer: 121-54-0 EINECS: 204-479-9 UN: 2923 ADR: 8,III Merck Index: 13,01074 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
Melting point | 158 to 163 °C |
Titration Argentometric | ≥96.0 % (on dry substance) |
Loss on drying | ≤5 % (105 °C, 4 hrs) |
Zinc (Zn) | ≤500 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...