4,4'-(1,2-Ethandiyl)dipyridin 97%

Lieferant: Thermo Fisher Scientific
Achtung

106470010 106470100
ACRO106470010EA 42 EUR
ACRO106470010 ACRO106470100
4,4'-(1,2-Ethandiyl)dipyridin 97%
1,2-Di(4-pyridyl)ethane

CAS No.: 4916-57-8

Formel: C₁₂H₁₂N₂
Schmelzpunkt: 107…113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006451
CAS-Nummer: 4916-57-8

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow to pink Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 107 to 113 °C
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR