(Ethoxycarbonylmethyl)triphenylphosphoniumbromid 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(Carbethoxymethyl)triphenylphosphonium bromide, (2-Ethoxy-2-oxoethyl)triphenylphosphonium bromide
108300250
ACRO108300250EA
34.9
EUR
ACRO108300250
(Ethoxycarbonylmethyl)triphenylphosphoniumbromid 98%
(Ethoxycarbonylmethyl)triphenylphosphoniumbromid
Formel:
C₂₂H₂₂BrO₂P Molecular Weight: 429,30 g/mol Schmelzpunkt: 158 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011835 CAS-Nummer: 1530-45-6 EINECS: 216-230-1 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥97.5 % |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...