N-Cbz-Glycin (Z-Gly-OH) 98.5%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-Carbobenzyloxyglycine, N-Benzyloxycarbonylglycine, Z-Gly-OH, 2-(((Benzyloxy)carbonyl)amino)acetic acid
108350250
108351000
108355000
ACRO108350250EA
35.6
EUR
ACRO108350250
ACRO108351000
ACRO108355000
N-Cbz-Glycin (Z-Gly-OH) 98.5%
N-Cbz-Glycin (Z-Gly-OH)
Formel:
C₁₀H₁₁NO₄ Molecular Weight: 209.2 g/mol Schmelzpunkt: 117…121 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002691 CAS-Nummer: 1138-80-3 EINECS: 214-516-0 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 117 to 122 °C |
HPLC | ≥98.0 % |
Titration with NaOH | ≥98.0 % |
Solubility | light yellow solution |
Titration with HClO4 | 97.5 to 102.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...