N-Benzyloxycarbonyl-L-Prolin 99%

Lieferant: Thermo Fisher Scientific

Synonyme: Z-Pro-OH, Z-L-Prolin, N-Cbz-L-proline, N-Carbobenzyloxy-L-proline, N-Benzyloxycarbonyl-L-proline

108370100 108370050
ACRO108370100EA 96.9 EUR
ACRO108370100 ACRO108370050
N-Benzyloxycarbonyl-L-Prolin 99%
N-Benzyloxycarbonyl-L-Prolin

CAS No.: 1148-11-4

Formel: C₁₃H₁₅NO₄
Molecular Weight: 249.27 g/mol
Schmelzpunkt: 76…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003170
CAS-Nummer: 1148-11-4

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
HPLC ≥98.5 %
Specific optical rotation -59° to -61° (20°C, 589 nm) (c=2, CH3COOH)
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR