Chelidamic acid monohydrate 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Oxo-1,4-dihydropyridine-2,6-dicarboxylic acid monohydrate

108470050
ACRO108470050EA 92.1 EUR
ACRO108470050
Chelidamic acid monohydrate 98%
Chelidamic acid monohydrate

Appearance: White to yellow Crystalline powder

Formel: C₇H₇NO₆
Molecular Weight: 201,134 g/mol
Schmelzpunkt: 263 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03818117
CAS-Nummer: 199926-39-1

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Water ≤10 %
Titration with NaOH ≥97.5 %
Phosphate (PO4) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤10 ppm
Free Iodine (I2) ≤0.75 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR