Chlordiphenylphosphin 95% tech.

Lieferant: Thermo Fisher Scientific
Gefahr

109130250 109131000 109135000
ACRO109130250EA 43.6 EUR
ACRO109130250 ACRO109131000 ACRO109135000
Chlordiphenylphosphin 95% tech.
Chlordiphenylphosphin
Formel: C₁₂H₁₀ClP
Siedepunkt: 320 °C (760 mmHg)
Schmelzpunkt: 14…16 °C
Dichte: 1,229 g/cm³
Flammpunkt: 138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00000529
CAS-Nummer: 1079-66-9
EINECS: 214-093-2
UN: 3265
ADR: 8,II

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Liquid (May develop some turbidity or precipitate)
Infrared spectrum Conforms
Refractive index 1.6340 to 1.6370 (20°C, 589 nm)
GC ≥94 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Carbonyl compounds ≤0.002 % (Acetone)
Titrable acid or base ≤0.0001 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR