2-(Chlormethyl)benzimidazol 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(Chlormethyl)-1H-benzimidazol

109390050 109390250
ACRO109390050EA 36.6 EUR
ACRO109390050 ACRO109390250
2-(Chlormethyl)benzimidazol 95%
2-(Chloromethyl)benzimidazole
Formel: C₈H₇ClN₂
Schmelzpunkt: 145…150 °C
MDL: MFCD00005602
CAS-Nummer: 4857-04-9
EINECS: 225-452-8

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to brown Powder
Infrared spectrum Conforms
Melting point 143 to 150 °C
HPLC ≥94 %
Titration with HClO4 94 to 106 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR