3-Chlorpyridin 99%

Lieferant: Thermo Fisher Scientific
Achtung

110020250 110021000
ACRO110020250EA 60.3 EUR
ACRO110020250 ACRO110021000
3-Chlorpyridin 99%
3-Chloropyridine
Formel: C₅H₄ClN
Siedepunkt: 147…150 °C (760 mmHg)
Schmelzpunkt: -0,5 °C
Dichte: 1,19 g/cm³
Flammpunkt: 66 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006375
CAS-Nummer: 626-60-8
EINECS: 210-955-7
ADR: 3,I

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5310 to 1.5340 (20°C, 589 nm)
GC ≥98.5 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 Passes test
Carbonyl compounds ≤0.001 % (each of acetone, formaldehyde, acetaldehyde)
Titrable acid ≤0.0003 meq/g
Titrable base ≤0.0002 meq/g
KMnO4-reducing substance Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR