L(+)-Citrullin 99%

Lieferant: Thermo Scientific

Synonyme: H-Cit-OH

110470250 110471000
ACRO110470250EA 144 EUR
ACRO110470250 ACRO110471000
L(+)-Citrullin 99%
L(+)-Citrullin
Formel: C₆H₁₃N₃O₃
Molecular Weight: 175,19 g/mol
Schmelzpunkt: 214 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064397
CAS-Nummer: 372-75-8
EINECS: 206-759-6

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.3 % (1 g, 105°C, 3 hrs)
Residue after ignition ≤0.1 %
UV transmittance ≥95.0 % (c=10, H2O, 430 nm) 1 cm cell
Specific optical rotation +24.5° to +26.8° (20°C, 589 nm) (c=8, 6N HCl)
Titration with HClO4 98.5 to 101.0 %
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤200 ppm
Ammonium (NH4) ≤200 ppm
Arsenic (As) ≤1 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR