3-Hydroxybenzonitril 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
m-Hydroxybenzonitril, 3-Cyanphenol, m-Cyanphenol
110820100
110820500
ACRO110820100EA
85.2
EUR
ACRO110820100
ACRO110820500
3-Hydroxybenzonitril 97%
3-Hydroxybenzonitril
Formel:
C₇H₅NO Molecular Weight: 119.12 g/mol Siedepunkt: 265 Schmelzpunkt: 75…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002252 CAS-Nummer: 873-62-1 EINECS: 212-845-4 UN: 3439 ADR: 6.1 III |
Spezifikation Testergebnisse
Appearance | White to light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 75 to 80 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...